6,6'-Di-tert-butyl-4,4'-thiodi-o-cresol ≥98.0% (by GC)
Toimittaja: TCI EUROPE
Varoitus
TCIAT1672-25GEA 51.7 EUR
TCIAT1672-25G
6,6'-Di-tert-butyl-4,4'-thiodi-o-cresol ≥98.0% (by GC)
6,6'-Di-tert-butyl-4,4'-thiodi-o-cresol
Kaava: C₂₂H₃₀O₂S
MDL numero: MFCD00008823
CAS-numero: 96-66-2
EINECS: 202-522-6
https://fi.vwr.com/stibo/web/std.lang.all/56/60/28995660.jpg
Order Now
Testitulosten erittely
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 128.0 deg-C