Powder containment cabinets, Excel Plus
Toimittaja: Bigneat
135-1907EA 5310 EUR
135-1907 135-1908 135-1910 135-1912 135-1911 135-1914 135-1913 135-1915
Powder containment cabinets, Excel Plus
Vetokaapit ja kotelot Vetokaapit
Excel Plus cabinets are for precision weighing applications - for protection from particles whilst weighing and handling powders. These cabinets incorporate aerofoil technology and graded rear back baffles ensuring reduced turbulence and precise control of air flow across the entire width of the working access. They are available as ducted or recirculatory models.
R models - recirculatory filtration
Fan speed is pre-set to between 0,3 and 0,5 m/s, depending on the application. Acrylic construction and angled clear front windows enable excellent vision of enclosed manipulations and comfort during use. The black base tray is ideal for highlighting spilt powder
Two-stage filtration is used in these particulate filtration systems. Cabinets are supplied complete with HEPA filtration (removes particles greater than 0,3 microns with an efficiency of 99,997%) and pre-filter (removes particles greater than 5 microns)
Keypad control panel with touch button controls and service due indicator
Gentle non-turbulent flow prevents balance fluctuation and cabinet dead spots
Suited to weighing with a 6 figure balance
Audible electronic low air flow alarm and lockable front access doors
Safe-change pre-filter can be bagged under negative pressure
Options include lighting (18 W fluorescent lamp), ionising bars to eliminate dust attraction and control of the static environment, an ULPA filtration upgrade and a static bench/stand (open or with cupboard doors). Please contact us for full details
The mild steel fan housing is epoxy coated. Transparent panels are manufactured from 12 mm clear cast acrylic which is non-flammable, resistant to corrosion and easy to clean. There is a full 175 mm height front access across full cabinet width. The base tray is manufactured from PVC
D models - ducted
Extraction of air can be through filters at a remote location or through existing extraction ductwork
Non-turbulent flow prevents balance fluctuation and cabinet dead spots
Angled clear front provides excellent vision of interior and comfort during use
Suited to weighing with a 6 figure balance
Electronic low airflow alarm
Transparent structure is manufactured from 12 mm clear cast acrylic which is non-flammable, resistant to corrosion and easy to clean. There is a full 175 mm height front access across full cabinet width. The base tray is manufactured from PVC
Options include lighting (18 W fluorescent lamp), ionising bars to eliminate dust attraction and control of the static environment, a spigot (200 mm) for connection to house ducting and a static bench/stand (open or with cupboard doors). Please contact us for full details