Acrylamide solutions, Sprint NEXT GEL®
M311-100MLEA 0 EUR
VWRCM311-100ML
Acrylamide solutions, Sprint NEXT GEL®
Pre-mixed, ready to pour acrylamide solution for the rapid analysis of standard SDS-Page mini-gels.
  • Ready-to-pour SDS polyacrylamide solutions - just add APS and TEMED
  • Faster gel casting with no stacking gel required
  • Broad range of separation
  • High resolution of protein bands
  • Faster electrophoresis time than standard NEXT GEL® solutions