Käsineet, teollisuus, nitriili, Nitrotough™ N650 & N660
Toimittaja: Ansell
Nitrotough™
112-2081EA 122 EUR
112-2081 MARG200FB0500 MARG200FB0508 112-2080 MARG200FB0509 MARG200FB0501
Käsineet, teollisuus, nitriili, Nitrotough™ N650 & N660
Käsineet
These cotton fleeced lined gloves with heavy duty supported nitrile finish are ideal for use in heavy mechanical handling, fabrication or site clearing.
  • Heavy duty coating
  • Fully coated to provide complete protection in a wet working environment
  • Excellent abrasion resistance and grip properties
  • Available with safety cuff or knitwrist cuff
Sertifikaatit: EN 388 (4221B); Cat. II