Käsineet, lämmönkestävät, Fireblade™ FB IV
Toimittaja: Marigold (Ansell)
Fireblade
MARGFB1V00000EA 60.11 EUR
MARGFB1V00000 MARGFB1V09000
Käsineet, lämmönkestävät, Fireblade™ FB IV
Käsineet
These protective gloves made of 100% Kevlar® over knitted cotton lining are suitable for all types of jobs requiring cut and thermal protection of the wrist and forearm.
  • High level of cut resistance for protection against sharp edges
  • Suitable for heat applications where protection is required up to 350 °C
  • Longer cuff protects wrist and forearm
  • Perfect for use in combination with Fireblade™ sleeves
Ideal for use in ceramic production, hot ovens, welding, brick and glass firing and processes involving steam production.
Sertifikaatit: EN 388 (254X), EN 407 (4341XX); Cat. II