Chemical resistant gloves, Orange Supaweight G02T
Toimittaja: Ansell
112-2693EA 0 EUR
MARG1614
Chemical resistant gloves, Orange Supaweight G02T
Unsupported natural rubber gloves with cotton flock lining for better comfort. The gloves feature a high resistance to water based chemicals, they are therefore ideal for laboratories and a broad range of chemical applications.
  • Roughened finish for excellent grip in damp conditions
  • Beaded cuff for tear resistance and easy donning
  • Extra length fluted cuff for ventilation and maximum protection
Length: 325 mm
Thickness: 1,35 mm (double wall)
Colour: Orange
EN 388 (2010), EN 374 (AKL); Cat. III; AQL < 0,65
Food contact approved