Büchner funnels
Toimittaja: Avignon
221-0125EA 61.1 EUR
221-0125 AVIG12709 AVIG12707 AVIG12708 AVIG12712 AVIG12710 AVIG12711 221-0123 221-0124
Büchner funnels
Made of white glazed porcelain.
  • Resistant to chemicals
  • Excellent temperature resistance
  • For paper or membrane filtration discs