Fall protection kit for roof work
Toimittaja: Miller by Honeywell
111-1373EA 309.7 EUR
DALL1002909
Fall protection kit for roof work
This ready to go kit comes in a plastic storage box and provides an immediate selection of fall protection equipment to suit the requirements of construction and especially roofing work. The set contains:
  • DuraFlex® elasticated harness, 3-point, with rear and front anchorage D-rings and PTFE finish
  • Rope lifeline, Ø 12 mm, length 15 m
  • Automatic rope grab, Ø 12 mm, with 0,3 m lanyard and hook
Sertifikaatit: EN 353