Huomiotakki, sade, palosuojattu, Marex 9464
Toimittaja: Sioen
Sio-Start® FR
113-7343EA 280 EUR
113-7343 113-7342 113-7334 113-7333 113-7344 113-7341 113-7340 113-7339 113-7336 113-7335 113-7338 113-7337
Huomiotakki, sade, palosuojattu, Marex 9464
Työvaatteet
This waterproof and windproof jacket is made of anti-static Sio-Start FR AST fabric: 100% polyester fabric with 100% FR PU coating + AST. Besides that, it features a fixed flame-retardant cotton lining and a detachable flame-retardant body fleece lining. The outer fabric is water-repellent, while the coating on the inside attracts moisture and keeps the skin dry.
  • High tear resistance, taped seams
  • Straight collar with foldaway hood
  • Zip closure under flap with touch & close fastening
  • Two patched pockets, Napoleon pocket, two loops for badges
  • Round inset sleeves, sleeve narrowing by press studs, elastic drawstring in hem
  • Additional reflective stripes over shoulders
Back length: 85 cm
Weight: 250 g/m²
Sertifikaatit: EN 531 (A B2 C1), EN 13034 (Type PB/6), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3), EN 471 (3-2)
Detachable lining only: EN 14058 (1XXXX)