1,3,5-Trithiane ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAT0532-25GEA
201
EUR
TCIAT0532-25G
1,3,5-Trithiane ≥98.0% (by GC)
1,3,5-Trithiane
Kaava:
C₃H₆S₃ |
MDL numero:
MFCD00006653 CAS-numero: 291-21-4 EINECS: 206-029-7 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 216.0 to 222.0 deg-C |
Solubility in hot Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...