6-Indoleboronic acid (contains varying amounts of Anhydride)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAI0803-1GEA
341
EUR
TCIAI0803-1G
6-Indoleboronic acid (contains varying amounts of Anhydride)
6-Indoleboronic acid
Kaava:
C₈H₈BNO₂ MW: 160,97 g/mol Sulamispiste: 200°C Storage Temperature: Jääkaappi |
MDL numero:
MFCD03095176 CAS-numero: 147621-18-9 |
Testitulosten erittely
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 95.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...