DL-Methionine sulphoxide ≥98.0% (by titrimetric analysis)
Toimittaja: TCI EUROPE
TCIAM0101-5GEA
66
EUR
TCIAM0101-5G
DL-Methionine sulphoxide ≥98.0% (by titrimetric analysis)
DL-Methionine sulphoxide
Kaava:
C₅H₁₁NO₃S MW: 165,21 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD00002620 CAS-numero: 62697-73-8 EINECS: 263-700-7 |
Testitulosten erittely
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...