Ethyl propyl sulphide ≥96.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAE0224-10GEA 103 EUR
TCIAE0224-10G
Ethyl propyl sulphide ≥96.0% (by GC)
Ethyl propyl sulphide
Kaava: C₅H₁₂S
Kiehumispiste: 117°C
MDL numero: MFCD00039940
CAS-numero: 4110-50-3
EINECS: 223-890-4
UN: 1993
ADR: 3,II
https://fi.vwr.com/stibo/web/std.lang.all/77/84/28967784.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR