Methyl octyl sulphide ≥98.0% (by GC)
Toimittaja: TCI EUROPE
TCIAM0379-25MLEA
129
EUR
TCIAM0379-25ML
Methyl octyl sulphide ≥98.0% (by GC)
Methyl octyl sulphide
Kaava:
C₉H₂₀S Kiehumispiste: 218°C |
MDL numero:
MFCD00039990 CAS-numero: 3698-95-1 EINECS: 223-029-2 |
![]() |
Testitulosten erittely
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...