Allyl methyl disulphide ≥90.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAA1939-5GEA 92.4 EUR
TCIAA1939-5G
Allyl methyl disulphide ≥90.0% (by GC)
Allyl methyl disulphide
Kaava: C₄H₈S₂
Leimahduspiste: 35°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00040023
CAS-numero: 2179-58-0
EINECS: 218-549-1
UN: 1993
ADR: 3,III
https://fi.vwr.com/stibo/web/std.lang.all/00/51/28930051.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR