4-Chloro-3-(ethoxycarbonyl)benzeneboronic acid, 96%
H52668.MDEA 96.4 EUR
ALFAH52668.MD ALFAH52668.03
4-Chloro-3-(ethoxycarbonyl)benzeneboronic acid, 96%
Purity: 96%
CAS No.: 874219-46-2
Molecular Formula: C9H10BClO4
Formula Weight: 228.44
Melting Point: 162-164°
MDL No.: MFCD08235062