4-(2,3-Dimethylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%

H54475.03EA 150 EUR
ALFAH54475.03 ALFAH54475.MD
4-(2,3-Dimethylphenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040197
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR