Benzeneseleninic acid ≥97.0% (by HPLC, titration analysis)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAB2134-5GEA
85.8
EUR
TCIAB2134-5G
Benzeneseleninic acid ≥97.0% (by HPLC, titration analysis)
Benzeneseleninic acid
Kaava:
C₆H₆O₂Se |
MDL numero:
MFCD00002100 CAS-numero: 6996-92-5 EINECS: 230-271-2 UN: 3283 ADR: 6.1,III |
Testitulosten erittely
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 121.0 to 125.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...