5-Benzyl(isopropyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
H54817.MDEA 90.9 EUR
ALFAH54817.MD ALFAH54817.03
5-Benzyl(isopropyl)carbamoyl-2-chlorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C17H19BClNO3
Formula Weight: 331.60
MDL No.: MFCD22125093