3-[Benzyl(methyl)carbamoyl]-4-fluorobenzeneboronic acid, 98%
H54918.03EA 726 EUR
ALFAH54918.03 ALFAH54918.MD
3-[Benzyl(methyl)carbamoyl]-4-fluorobenzeneboronic acid, 98%
Purity: 98%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20040270