4'-(4-Bromophenyl)acetophenone ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAA1410-25GEA
106
EUR
TCIAA1410-25G
4'-(4-Bromophenyl)acetophenone ≥98.0% (by GC)
4'-(4-Bromophenyl)acetophenone
Kaava:
C₁₄H₁₁BrO MW: 275,15 g/mol |
MDL numero:
MFCD00143242 CAS-numero: 5731-01-1 EINECS: 227-236-9 |
Testitulosten erittely
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...