Ethyl perfluorononanoate ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAH1038-5GEA
97.9
EUR
TCIAH1038-5G
Ethyl perfluorononanoate ≥98.0% (by GC)
Ethyl perfluorononanoate
Kaava:
C₁₁H₅F₁₇O₂ |
MDL numero:
MFCD00039245 CAS-numero: 30377-52-7 EINECS: 250-159-7 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...