Methyl perfluorononanoate ≥95.0% (by GC)

Toimittaja: TCI EUROPE

TCIAM1915-25GEA 160 EUR
TCIAM1915-25G
Methyl perfluorononanoate ≥95.0% (by GC)
Methyl perfluorononanoate
Kaava: C₁₀H₃F₁₇O₂
MW: 478,11 g/mol
Kiehumispiste: 65°C
MDL numero: MFCD00153213
CAS-numero: 51502-45-5
https://fi.vwr.com/stibo/web/std.lang.all/78/60/28987860.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR