3',5'-Dibenzyloxyacetophenone ≥97.0% (by GC)
Toimittaja: TCI EUROPE
TCIAD2085-10GEA
180
EUR
TCIAD2085-10G
3',5'-Dibenzyloxyacetophenone ≥97.0% (by GC)
3',5'-Dibenzyloxyacetophenone
Kaava:
C₂₂H₂₀O₃ MW: 332,40 g/mol |
MDL numero:
MFCD00004777 CAS-numero: 28924-21-2 EINECS: 249-315-7 |
Testitulosten erittely
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 61.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...