5-Methylnonane ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAM0284-5MLEA 460 EUR
TCIAM0284-5ML
5-Methylnonane ≥98.0% (by GC)
5-Methylnonane
Kaava: C₁₀H₂₂
Kiehumispiste: 165°C
Leimahduspiste: 38°C
MDL numero: MFCD00048832
CAS-numero: 15869-85-9
UN: 3295
ADR: 3,III
https://fi.vwr.com/stibo/web/std.lang.all/36/94/28973694.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR