1,3-Dibromo-5-(trifluoromethoxy)benzene ≥98.0% (by GC)
Toimittaja: TCI EUROPE
Varoitus
TCIAD3322-5GEA 56.1 EUR
TCIAD3322-5G TCIAD3322-25G
1,3-Dibromo-5-(trifluoromethoxy)benzene ≥98.0% (by GC)
1,3-Dibromo-5-(trifluoromethoxy)benzene
Kaava: C₇H₃Br₂F₃O
MW: 319,90
MDL numero: MFCD00075280
CAS-numero: 207226-31-1
https://fi.vwr.com/stibo/web/std.lang.all/03/01/28980301.jpg
Order Now
Testitulosten erittely
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %