4'-Hydroxyhexanophenone ≥99.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAH0884-5GEA 72.6 EUR
TCIAH0884-5G
4'-Hydroxyhexanophenone ≥99.0% (by GC)
4'-Hydroxyhexanophenone
Kaava: C₁₂H₁₆O₂
Kiehumispiste: 167°C
MDL numero: MFCD00671540
CAS-numero: 2589-72-2
https://fi.vwr.com/stibo/web/std.lang.all/09/06/28970906.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 62.0 to 65.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR