New fuchsin, Magenta III
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAN0478-25GEA
38.5
EUR
TCIAN0478-25G
New fuchsin, Magenta III
New fuchsin, Magenta III
Kaava:
C₂₂H₂₄ClN₃ MW: 365,91 g/mol |
MDL numero:
MFCD00012567 CAS-numero: 3248-91-7 EINECS: 221-831-7 |
Testitulosten erittely
Appearance | Green to Dark green powder to crystaline |
Purity (HPLC) | min. 85.0 area% |
Lambda max. | 550.0 to 558.0 nm (EtOH) |
Absorbance (E1%1cm) | min. 1,500 (EtOH, 550.0 to 558.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...