Käsineet, tutkimus, fluoresoivat, Showa N-Dex® 9500PF
Toimittaja: Showa
112-3289EA 33.3 EUR
BEST9500PFM BEST9500PFXS BEST9500PFL BEST9500PFXL
Käsineet, tutkimus, fluoresoivat, Showa N-Dex® 9500PF
These ambidextrous high visibility gloves protect the hand from a wide array of chemical hazards while providing increased safety in poor lighting conditions. Ideal for light assembly tasks in chemical and medical industries.
  • Non sterile, powder-free, smooth finish
  • 100% accelerator-free, contain no latex
  • Feel like a second skin
  • Cuff prevents dirt from entering the glove; easy to put on and remove
  • Easy to tear away in case of emergency break
  • Great freedom of movement for the wrist
Ideal for light assembly tasks in chemical and medical industries.
Sertifikaatit: EN 455; Cat. I