ESD-työkengät, nauhoilla, Business Men, 32922
Toimittaja: Abeba
Business Men
114-3767EA 116 EUR
ABEB32922/43 ABEB32922/45 ABEB32922/44 ABEB32922/46 ABEB32922/42 ABEB32922/47 ABEB32922/39 ABEB32922/41 ABEB32922/40
ESD-työkengät, nauhoilla, Business Men, 32922
ESD business shoe made from smooth leather with slip resistant sole, calf leather lining and leather insole.
  • Instep area with lacing
  • Slip resistant special rubber sole
Colour: Black
Sertifikaatit: EN 61340 (ESD); OB, A, SRA