Ion exchange chromatography media, Capto™ ViralQ
Toimittaja: Cytiva
28-9032-31EA 0 EUR
GEHE28-9032-31
Ion exchange chromatography media, Capto™ ViralQ
Capto™ Q is a strong anion exchange bioprocessing medium for capture and intermediate purification of proteins from large feed volumes by packed bed chromatography. Capto™ ViralQ is in all respects identical to Capto™ Q, but in addition it provides a license to be used for virus purification.
  • Raised productivity with high dynamic binding capacity at high flow
  • Increased yield with rapid mass transfer
  • Reduced process time with high volume throughput
  • Cost-effective processing with smaller unit operations
  • Possibilities to perform effective and rigorous CIP procedures