Affinity chromatography media, DNA cellulose
Toimittaja: Cytiva
27-5579-02EA 259 EUR
GEHE27-5579-02
Affinity chromatography media, DNA cellulose
Immobilised calf thymus DNA media is used for the affinity isolation of proteins that bind DNA.