N-Phenylurethane ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAP0165-500GEA 182 EUR
TCIAP0165-500G TCIAP0165-25G
N-Phenylurethane ≥98.0% (by GC)
N-Phenylurethane
Kaava: C₉H₁₁NO₂
MW: 165,19
Kiehumispiste: 237°C
MDL numero: MFCD00026806
CAS-numero: 101-99-5
EINECS: 202-995-9
https://fi.vwr.com/stibo/web/std.lang.all/08/94/28990894.jpg

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR