[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex (1:1) (13% Pd)

Toimittaja: Thermo Scientific
Varoitus

41225.14EA 2270 EUR
41225.14 41225.03 41225.06
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex (1:1) (13% Pd)
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Kaava: C₃₅H₃₀Cl₄FeP₂Pd
MW: 816,64 g/mol
Sulamispiste: 275…280 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00792899
CAS-numero: 95464-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR