D-(+)-Glucurono-6,3-lactone

Toimittaja: Molekula

MOLE13139248-100GEA 107 EUR
MOLE13139248-100G
D-(+)-Glucurono-6,3-lactone
D-(+)-Glucurono-6,3-lactone
Kaava: C₆H₈O₆
MW: 176,12 g/mol
MDL numero: MFCD00135622
CAS-numero: 32449-92-6
EINECS: 251-053-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR