Retinoic acid

Toimittaja: Molekula

MOLEM26143112EA 113 EUR
MOLEM26143112
Retinoic acid
Retinoic acid
Kaava: C₂₀H₂₈O₂
MDL numero: MFCD00001551
CAS-numero: 302-79-4
EINECS: 206-129-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR