1-Naphthylacetamide

Toimittaja: EHRENSTORFER GMBH

EHERC15450000EA 86.1 EUR
EHERC15450000
1-Naphthylacetamide
1-Naphthylacetamide
Kaava: C₁₂H₁₁NO
MW: 185,23 g/mol
MDL numero: MFCD00004047
CAS-numero: 86-86-2
EINECS: 201-704-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR