4-Boronobenzenesulphonamide

Toimittaja: Apollo Scientific

APOSOR1186-5GEA 519 EUR
APOSOR1186-5G APOSOR1186-250MG
4-Boronobenzenesulphonamide
4-Boronobenzenesulphonamide
Kaava: C₆H₈BNO₄S
MW: 201,00
MDL numero: MFCD06659839
CAS-numero: 613660-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR