2-(4-Chloro-3-sulphamoylbenzoyl)benzoic acid

Toimittaja: U.S. Pharmacopeia

USPH1119309EA 1130 EUR
USPH1119309
2-(4-Chloro-3-sulphamoylbenzoyl)benzoic acid
2-(4-Chloro-3-sulphamoylbenzoyl)benzoic acid
Kaava: C₁₄H₁₀ClNO₅S
CAS-numero: 5270-74-6
EINECS: 226-089-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR