Dichloromethyloctylsilane ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAO0265-25MLEA 36.3 EUR
TCIAO0265-25ML
Dichloromethyloctylsilane ≥97.0% (by GC)
Dichloromethyloctylsilane
Kaava: C₉H₂₀Cl₂Si
MW: 227,25 g/mol
Kiehumispiste: 234°C
Leimahduspiste: 43°C
MDL numero: MFCD00042606
CAS-numero: 14799-93-0
EINECS: 238-863-2
UN: 2986
ADR: 8,II
https://fi.vwr.com/stibo/web/std.lang.all/05/36/28990536.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR