4'-Aminohexanophenone ≥98.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAA2058-25GEA
571
EUR
TCIAA2058-25G
TCIAA2058-5G
4'-Aminohexanophenone ≥98.0% (by GC, titration analysis)
4'-Aminohexanophenone
Kaava:
C₁₂H₁₇NO |
CAS-numero:
38237-76-2 UN: 2811 ADR: 6.1,III |
![]() |
Testitulosten erittely
Appearance | White to Light red to Green powder to crystalline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...