3-Chloropropyl chloroformate ≥95.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAC1983-25GEA 91.3 EUR
TCIAC1983-25G
3-Chloropropyl chloroformate ≥95.0% (by GC)
3-Chloropropyl chloroformate
Kaava: C₄H₆Cl₂O₂
Leimahduspiste: 96°C
MDL numero: MFCD02093717
CAS-numero: 628-11-5
EINECS: 425-770-9
UN: 3277
ADR: 6.1,II
https://fi.vwr.com/stibo/web/std.lang.all/90/53/28939053.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR