Octyl chloroformate ≥97.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAC0724-25MLEA
161
EUR
TCIAC0724-25ML
Octyl chloroformate ≥97.0% (by GC, titration analysis)
Octyl chloroformate
Kaava:
C₉H₁₇ClO₂ Kiehumispiste: 123°C Leimahduspiste: 76°C |
MDL numero:
MFCD00000652 CAS-numero: 7452-59-7 EINECS: 231-224-9 UN: 3277 ADR: 6.1,II |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...