D-(+)-Fucose ≥98.0% (by HPLC)
Toimittaja: TCI EUROPE
TCIAD0049-1GEA
96.8
EUR
TCIAD0049-1G
TCIAD0049-5G
D-(+)-Fucose ≥98.0% (by HPLC)
D-(+)-Fucose
Kaava:
C₆H₁₂O₅ |
MDL numero:
MFCD00135603 CAS-numero: 3615-37-0 EINECS: 222-792-9 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Specific rotation [a]20/D | +72.0 to +77.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...