2,3',4,4'-Tetrahydroxybenzophenone ≥90.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAT1371-5GEA
180
EUR
TCIAT1371-5G
2,3',4,4'-Tetrahydroxybenzophenone ≥90.0% (by GC, titration analysis)
2,3',4,4'-Tetrahydroxybenzophenone
Kaava:
C₁₃H₁₀O₅ |
MDL numero:
MFCD00142775 CAS-numero: 61445-50-9 |
Testitulosten erittely
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 90.0 % |
Purity (Neutralization titration) | min. 90.0 % |
Melting point | 200.0 to 204.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...