3'-Acetoxyacetophenone ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAA1996-5GEA 39.6 EUR
TCIAA1996-5G TCIAA1996-25G
3'-Acetoxyacetophenone ≥98.0% (by GC)
3'-Acetoxyacetophenone
Kaava: C₁₀H₁₀O₃
Kiehumispiste: 147°C
MDL numero: MFCD00017227
CAS-numero: 2454-35-5
EINECS: 219-524-8
https://fi.vwr.com/stibo/web/std.lang.all/01/37/28930137.jpg

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR