Pentadecanophenone ≥98.0% (by GC)
Toimittaja: TCI EUROPE
TCIAP1903-5GEA
167
EUR
TCIAP1903-5G
Pentadecanophenone ≥98.0% (by GC)
Pentadecanophenone
Kaava:
C₂₁H₃₄O |
CAS-numero:
4669-04-9 |
Testitulosten erittely
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...