4'-Butoxyacetophenone ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAB1501-5GEA
67.1
EUR
TCIAB1501-5G
4'-Butoxyacetophenone ≥98.0% (by GC)
4'-Butoxyacetophenone
Kaava:
C₁₂H₁₆O₂ MW: 192,26 g/mol |
MDL numero:
MFCD00027200 CAS-numero: 5736-89-0 EINECS: 227-248-4 |
![]() |
Testitulosten erittely
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Freezing point | 21.0 to 24.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...