2-tert-Butyltoluene ≥99.0% (by GC)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAB0966-1MLEA
160
EUR
TCIAB0966-1ML
2-tert-Butyltoluene ≥99.0% (by GC)
2-tert-Butyltoluene
Kaava:
C₁₁H₁₆ MW: 148,25 g/mol Kiehumispiste: 200°C |
MDL numero:
MFCD00059209 CAS-numero: 1074-92-6 UN: 2667 ADR: 6.1,III |
Testitulosten erittely
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...