(Methoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAM1326-25GEA
60.5
EUR
TCIAM1326-25G
(Methoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
(Methoxycarbonylmethyl)triphenylphosphonium bromide
Kaava:
C₂₁H₂₀BrO₂P MW: 415,28 g/mol |
MDL numero:
MFCD00011801 CAS-numero: 1779-58-4 EINECS: 217-222-0 |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Precipitation Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...